EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Inhibition of autophagy sensitises cells to hydrogen peroxide-induced apoptosis: Protective effect of mild thermotolerance acquired at 40°C.
PMID 27666506
Abstract
Various toxic compounds produce reactive oxygen species, resulting in oxidative stress that threatens cellular homeostasis. Yet, lower doses of stress can stimulate defence systems allowing cell survival, whereas intense stress activates cell death pathways such as apoptosis. Mild thermal stress (40°C, 3h) induces thermotolerance, an adaptive survival response that renders cells less sensitive to subsequent toxic stress, by activating defence systems like heat shock proteins, antioxidants, anti-apoptotic and ER-stress factors. This study aims to understand how autophagy and apoptosis are regulated in response to different doses of H
Related Materials