EMAIL THIS PAGE TO A FRIEND
Artificial cells, nanomedicine, and biotechnology
Plant phenyl-propanoids-conjugated silver nanoparticles from edible plant Suaeda maritima (L.) dumort. Inhibit proliferation of K562-human myeloid leukemia cells.
PMID 27677829
Abstract
The present study portrays the isolation of four phenylpropanoids - ferulic acid (FA), sinapic acid (SA), caffeic acid (CA), and chlorogenic acid (CHA) from the water extract of Suaeda maritima (L.) Dumort, a phytochemically less explored Indian medicinal plant. Further, synthesis and characterization of silver and gold nanoparticles using the isolated phenylpropanoids were done. The silver nanoparticles synthesized from S. maritima water extract along with silver nano-conjugated forms of the isolated compounds exhibited appreciable anti-leukemic activity against K562 cells (human myeloid leukemia). Especially, the ferulic and CA-conjugated silver nanoparticles showed significant (P < .01) activity against leukemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
132292
2-Vinylpyridine, 97%
C7H7N