EMAIL THIS PAGE TO A FRIEND
Journal of translational medicine
The fibrolytic potentials of vitamin D and thymoquinone remedial therapies: insights from liver fibrosis established by CCl4 in rats.
PMID 27681697
Abstract
Vitamin D (VitD) and thymoquinone (TQ) are nutraceutical agents with well-known immunomodulatory and hepatoprotective properties. This study measured whether VitD and TQ, individually or combined, could have direct fibrolytic activities and/or enhanced performance during remedial treatment of liver fibrosis established by CCl4 in rats. Eighty five male Wistar rats were used as 10 negative controls (NC) and the remainders were distributed equally into 5 groups: short (S-PC) and long (L-PC) positive controls, TQ, VitD and VitD/TQ groups. CCl4 was injected for 7 weeks followed by a week of no intervention. TQ and/or VitD were given orally (3 days/week) from week 9 and euthanasia was at week 17 for all groups except the S-PC was at week 9. Following histopathological and digital image analyses, TGF-β1, IL-6, IL-10, IL-22 and MMP-9 were measured by ELISA in liver homogenates while the corresponding cytokine receptors were measured by immunohistochemistry. The mRNA expressions of all molecules were measured by quantitative RT-PCR. Fibrosis was evident in both PC-groups and was significantly more advanced in the L-PC than S-PC, reaching to cirrhosis. The concentrations of TGF-β1, IL-6, IL-22 and their receptors were significantly higher (P < 0.05) simultaneously with significantly lower (P < 0.05) concentrations of MMP-9, IL-10 and IL-10 receptors in the S-PC and L-PC than the NC-group. TQ and VitD monotherapies showed significantly less fibrosis than L-PC but were similar to S-PC. Both remedial monotherapies also resulted in significant decreases of TGF-β1, IL-6, IL-22 and their receptors together with significant increases of MMP-9 and IL-10 system compared with S-PC and L-PC groups. Interestingly, dual therapy resulted in the most significant improvement in fibrosis score and index, yet was significantly higher (P < 0.05) than the NC-group, and concurred with the utmost significant restorations of all candidate genes and proteins. VitD and TQ exhibited comparable anti-fibrogenic effects and modulated several pro- and anti-fibrotic mediators. Additionally, VitD/TQ dual therapy alleviated the previously established liver fibrosis simultaneously with significantly enhanced actions at the molecular level. More studies are required to explorer the therapeutic value of TQ and VitD against liver fibrosis in human.