Analytical and bioanalytical chemistry
A non-enzymatic urine glucose sensor with 2-D photonic crystal hydrogel.
PMID 27682838
A novel polymerized crystalline colloidal array (PCCA) sensing material for the detection of urine glucose was developed by embedding a two-dimensional (2-D) polystyrene crystalline colloidal array (CCA) in 3-acrylamidophenylboronic acid (3-APBA)-functionalized hydrogel. After adjusting the cross-linker concentration, this material showed significant sensitivity for glucose under lab conditions, the particle spacing of the PCCA changed from 917 to 824xa0nm (93xa0nm) within 3xa0min as the glucose concentration increased from 0 to 10xa0mM, and the structural color of the PCCA changed from red through orange, to green, and finally, to cyan. In further experiments, this material was used to semi-quantitatively detect glucose in 20 human urine (HU) samples. Compared with the traditional dry-chemistry method, which was applied widely in clinical diagnosis, the PCCA method was more accurate and cost-effective. Moreover, this method can efficiently avoid the errors induced by most of the urine-interfering elements like vitamin C and ketone body. With a homemade portable optical detector, this low-cost intelligent sensing material can provide a more convenient and efficient strategy for the urine glucose detection in clinical diagnosis and point-of-care monitoring.