EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Extracellular Loop 2 of the Adenosine A1 Receptor Has a Key Role in Orthosteric Ligand Affinity and Agonist Efficacy.
PMID 27683014
Abstract
The adenosine A