EMAIL THIS PAGE TO A FRIEND
Thrombosis and haemostasis
Enhanced platelet MRP4 expression and correlation with platelet function in patients under chronic aspirin treatment.
PMID 27683757
Abstract
Platelet multidrug resistance protein4 (MRP4)-overexpression has a role in reducing aspirin action. Aspirin in vivo treatment enhances platelet MRP4 expression and MRP4 mediated transport inhibition reduces platelet function and delays thrombus formation. The aim of our work was to verify whether MRP4 expression is enhanced in platelets obtained from patients under chronic aspirin treatment and whether it correlates with residual platelet reactivity. We evaluated changes on mRNA and protein-MRP4 expression and platelet aggregation in four populations: healthy volunteers (HV), aspirin-free control population (CTR), patients who started the treatment less than one month ago (ASA<1 month patients) and aspirinated patients who started the treatment more than two months ago (ASA>2 months patients). In platelets obtained from ASA>2 months patients, it was found a statistically significant MRP4 enhancement of both mRNA and protein expression compared to HV, CTR and ASA<1 month patients. Platelets obtained from ASA>2 months patients that present high levels of platelet MRP4, have higher serum TxB