EMAIL THIS PAGE TO A FRIEND
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology
Characterization of the functional and anatomical differences in the atrial and ventricular myocardium from three species of elasmobranch fishes: smooth dogfish (Mustelus canis), sandbar shark (Carcharhinus plumbeus), and clearnose skate (Raja eglanteria).
PMID 27686667
Abstract
We assessed the functional properties in atrial and ventricular myocardium (using isolated cardiac strips) of smooth dogfish (Mustelus canis), clearnose skate (Raja eglanteria), and sandbar shark (Carcharhinus plumbeus) by blocking Ca