EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Structure and function of anhydride-modified forms of human insulin: In silico, in vitro and in vivo studies.
PMID 27687638
Abstract
Insulin is a therapeutic protein whose amyloid formation is reported in diabetic patients. Four anhydride compounds were used in the current study in order to experiment their potential reducing effect on insulin propensity to form amyloid fibrils. The modified forms (obtained with succinic-, 3,3-dimethylglutaric, 2-phenylglutaric-, and (2-Dodecen-1-yl) succinic anhydride), were first characterized with regard to melting temperature (Tm), changes in secondary structure percentage and hydrophobic surface. Fibril formation was then assessed by Congo red absorbance kinetics and transmission electron microscopy. Functionality was investigated with the use of an insulin tolerance test in NMRI mice. Finally, 10ns molecular dynamics simulations were performed during which structural changes, potential energy, gyration radius, RMSD, and accessible surface area were monitored. In all cases, α-helical structure content of the modified forms was reduced, but thermal stability and structural compactness of modified insulin were increased except in case of the dodecenylated species. All modified insulin forms undergo amorphous aggregation instead of amyloid fibrils formation, and dodecenylated insulin makes the largest amorphous aggregates. In silico results were overall in accordance with in vitro studies. Finally, only succinylated insulin was functional, although dimethylglutaric-modified insulin started to show some activity after 2h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3516
Thioflavin T, used as stain for amyloid
C17H19ClN2S