EMAIL THIS PAGE TO A FRIEND
Clinical and experimental reproductive medicine
The effect of the site of laser zona opening on the complete hatching of mouse blastocysts and their cell numbers.
PMID 27689037
Abstract
We studied the effect of the site of laser zona opening on the complete hatching of mouse blastocysts and the cell numbers of the completely hatched blastocysts. Mouse blastocysts were randomly allocated to the inner cell mass (ICM) group (zona opening performed at the site of the ICM, n=125), the trophectoderm (TE) group (zona opening performed opposite to the ICM, n=125) and the control group (no zona opening, n=125). The rate of complete hatching of the blastocysts was not significantly different in the ICM and the TE group (84.8% vs 80.8%, respectively; p=0.402), but was significantly lower in the control group (51.2%, p<0.001). The cell numbers in the completely hatched blastocysts were comparable in the control group, the ICM group, and the TE group (69±19.3, 74±15.7, and 71±16.8, respectively; p=0.680). These findings indicate that the site of laser zona opening did not influence the rate of complete hatching of mouse blastocysts or their cell numbers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5774 Anti-Mouse Serum antibody produced in rabbit, whole antiserum