EMAIL THIS PAGE TO A FRIEND
Aging
Clinical significance and prognostic value of TRIM24 expression in esophageal squamous cell carcinoma.
PMID 27689360
Abstract
Tripartite motif-containing 24 (TRIM24), a member of the transcription intermediary factor 1 family, is defined as a co-regulator with several nuclear receptors, such as RARα. TRIM24 has been reported to be involved in many cancers. In this study, we aimed to investigate the expression pattern and prognostic significance of TRIM24 and its relationship with RARα in esophageal squamous cell cancer (ESCC). Both mRNA and protein expression levels of TRIM24 were found to be significantly decreased in ESCC, as judged by qRT-PCR and western blot. Immunohistochemistry staining shows that the reduced TRIM24 protein is associated with lymph node metastasis (