EMAIL THIS PAGE TO A FRIEND
Journal of biomedical materials research. Part B, Applied biomaterials
Substitutional limit of gadolinium in β-tricalcium phosphate and its magnetic resonance imaging characteristics.
PMID 27690186
Abstract
To compensate the limitations of bone tissue magnetic resonance imaging (MRI), a series of gadolinium (Gd