EMAIL THIS PAGE TO A FRIEND
Oncotarget
Tyrosine kinase LYN is an oncotarget in human cervical cancer: A quantitative proteomic based study.
PMID 27690342
Abstract
Cervical cancer is one of the most common malignant tumor in women. The mechanisms of cervical cancer are intricate and have not been fully understood. Therefore, we employed iTRAQ to obtain novel proteins profile which participates in the tumor oncogenesis of cervical cancer. 3300 proteins were identified aberrantly expressed in cervical cancer, and western bolt was performed to validate the results of iTRAQ. Then, we selected LYN for further study. Immunohistochemistry identified that LYN expression was significantly increased in cervical cancer tissues than that in cancer adjacent normal cervical tissues and normal cervical tissues. The increased LYN expression was significantly correlated with cancer differentiation and FIGO stage. Silencing LYN inhibited cell proliferation, migration and invasion, conversely, overexpression LYN promoted cell proliferation, migration and invasion. In terms of mechanism, LYN could also promote cervical cancer cells metastasis through activating IL-6/STAT3 pathway. In vivo study, overexpression LYN promoted tumor growth, meanwhile knockdown LYN inhibited tumor growth. These results indicate that LYN tyrosine kinase is an oncogenic gene and can serve as a novel target for cervical cancer research and therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU089961 MISSION® esiRNA, esiRNA human LYN (esiRNA1)