Ultrasound in medicine & biology
Topical Fish Oil Application Coupling with Therapeutic Ultrasound Improves Tendon Healing.
PMID 27692307
Fish oil has been shown to promote collagen synthesis, and hence, connective tissue healing. Therapeutic ultrasound is commonly used to treat soft tissue injuries. This study aimed to investigate the therapeutic effect of topical fish oil on the management of Achilles tendon rupture, comparing normal therapeutic ultrasound with a combination of ultrasound and fish oil. Eighty-five Sprague-Dawley rats underwent surgical hemitenotomy of the right medial Achilles tendon. The rats received daily treatment of either topical placebo ointment (control group [CON]), topical fish oil (FO), therapeutic ultrasound (US) or ultrasound with fish oil as the coupling medium (FU). The treatment started on post-surgical day 2 over a 2-wk or 4-wk period. On days 15 and 29, the rats were sacrificed and their Achilles tendons were tested for structural stiffness, ultimate tensile strength (UTS) and energy absorption capacity. At 2xa0wk, only US showed higher normalized UTS compared with CON (pxa0
Related Materials
Product #
Molecular Formula
Add to Cart
F8020 Fish oil from menhaden, crude source of omega-3 fatty acids