EMAIL THIS PAGE TO A FRIEND
Journal of assisted reproduction and genetics
Blastomere biopsy for PGD delays embryo compaction and blastulation: a time-lapse microscopic analysis.
PMID 27696105
Abstract
The purpose of the study was to explore the effect of blastomere biopsy for preimplantation genetic diagnosis (PGD) on the embryos' dynamics, further cleavage, development, and implantation. The study group included 366 embryos from all PGD treatments (September 2012 to June 2014) cultured in the EmbryoScope™ time-lapse monitoring system. The control group included all intracytoplasmic sperm injection (ICSI) embryos cultured in EmbryoScope™ until day 5 during the same time period (385 embryos). Time points of key embryonic events were analyzed with an EmbryoViewer™. Most (88 %) of the embryos were biopsied at ≥8 cells. These results summarize the further dynamic development of the largest cohort of biopsied embryos and demonstrate that blastomere biopsy of cleavage-stage embryos significantly delayed compaction and blastulation compared to the control non-biopsied embryos. This delay in preimplanation developmental events also affected postimplantation development as observed when the dynamics of non-implanted embryos (known implantation data (KID) negative) were compared to those of implanted embryos (KID positive). Analysis of morphokinetic parameters enabled us to explore how blastomere biopsy interferes with the dynamic sequence of developmental events. Our results show that biopsy delays the compaction and the blastulation of the embryos, leading to a decrease in implantation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5249
Cetrorelix acetate, ≥98% (HPLC)
C70H92ClN17O14 · xC2H4O2