EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
TFP5 prevents 1-methyl-4-phenyl pyridine ion-induced neurotoxicity in mouse cortical neurons.
PMID 27698762
Abstract
The present study aimed to investigate the protective effect of a modified p5 peptide, TFP5, on 1-methyl-4-phenyl pyridine ion (MPP
Related Materials