EMAIL THIS PAGE TO A FRIEND
In vitro cellular & developmental biology. Animal
Insulin-like growth factor 1 regulation of proliferation and differentiation of Xenopus laevis myogenic cells in vitro.
PMID 27699652
Abstract
To understand the mechanism of muscle remodeling during Xenopus laevis metamorphosis, we examined the in vitro effect of insulin-like growth factor 1 (IGF-1) on growth and differentiation of three different-fate myogenic cell populations: tadpole tail, tadpole dorsal, and young adult leg muscle. IGF-1 promoted growth and differentiation of both tail and leg myogenic cells only under conditions where these cells could proliferate. Inhibition of cell proliferation by DNA synthesis inhibitor cytosine arabinoside completely canceled the IGF-1's cell differentiation promotion, suggesting the possibility that IGF-1's differentiation-promotion effect is an indirect effect via IGF-1's cell proliferation promotion. IGF-1 promoted differentiation dose dependently with maximum effect at 100-500 ng/ml. RT-PCR analysis revealed the upregulation (11-fold) of ifg1 mRNA expression in developing limbs, suggesting that IGF-1 plays a role in promoting muscle differentiation during limb development. The combined effect of triiodo-L-thyronine (T
Related Materials