EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Increased Amyloid Precursor Protein and Tau Expression Manifests as Key Secondary Cell Death in Chronic Traumatic Brain Injury.
PMID 27699791
Abstract
In testing the hypothesis of Alzheimer's disease (AD)-like pathology in late stage traumatic brain injury (TBI), we evaluated AD pathological markers in late stage TBI model. Sprague-Dawley male rats were subjected to moderate controlled cortical impact (CCI) injury, and 6 months later euthanized and brain tissues harvested. Results from H&E staining revealed significant 33% and 10% reduction in the ipsilateral and contralateral hippocampal CA3 interneurons, increased MHCII-activated inflammatory cells in many gray matter (8-20-fold increase) and white matter (6-30-fold increased) regions of both the ipsilateral and contralateral hemispheres, decreased cell cycle regulating protein marker by 1.6- and 1-fold in the SVZ and a 2.3- and 1.5-fold reductions in the ipsilateral and contralateral dentate gyrus, diminution of immature neuronal marker by two- and onefold in both the ipsilateral and contralateral SVZ and dentate gyrus, and amplified amyloid precursor protein (APP) distribution volumes in white matter including corpus callosum, fornix, and internal capsule (4-38-fold increase), as well as in the cortical gray matter, such as the striatum hilus, SVZ, and dentate gyrus (6-40-fold increase) in TBI animals compared to controls (P's < 0.001). Surrogate AD-like phenotypic markers revealed a significant accumulation of phosphorylated tau (AT8) and oligomeric tau (T22) within the neuronal cell bodies in ipsilateral and contralateral cortex, and dentate gyrus relative to sham control, further supporting the rampant neurodegenerative pathology in TBI secondary cell death. These findings indicate that AD-like pathological features may prove to be valuable markers and therapeutic targets for late stage TBI. J. Cell. Physiol. 232: 665-677, 2017. © 2016 Wiley Periodicals, Inc.