EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Bioactive potential and structural chracterization of sulfated polysaccharide from seaweed (Gracilaria corticata).
PMID 27702543
Abstract
The Sulfated polysaccharide was purified through anion-exchange and gel permeation column chromatography. The isolated sulfated polysaccharide from C. corticata contains 84% of carbohydrate, 0% of protein, 19.7% of ash and 29.4% of moisture was found. The carbon, hydrogen, nitrogen and sulfur content as 33.19%, 5.91%, 7.21% and 3.75%. The molecular weight of sulfated polysaccharide was found to be 43kDa. The sugar was composed of (90.11%), glucose (5.47%), xylose (2.30%) and mannose (2.12%). The structural feature of sulfated polysachharide was studied through FT-IR and
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W373011 L-Rhamnose, natural sourced, 99%, FG
C6H12O5H2O