EMAIL THIS PAGE TO A FRIEND
Chemosphere
A passive sampling method for assessing the occurrence and risk of organophosphate flame retardants in aquatic environments.
PMID 27705807
Abstract
A modified polar organic chemical integrative sampler (m-POCIS) was used to determine the occurrence of and risk posed by organophosphate flame retardants (OPFRs) in the Yangtze River in Nanjing. Laboratory calibrations were performed to determine sampling rates (R