EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and topoisomerases inhibitory activity of heteroaromatic chalcones.
PMID 27707625
Abstract
The critical role of nuclear topoisomerase enzymes during cell proliferation process guided topoisomerases to be one of the major targets for anticancer drug development. We have designed and synthesized 22 heteroaromatic ring incorporated chalcone derivatives substituted with epoxide or thioepoxide. Topoisomerase enzyme inhibitory activity and cytotoxic tests were also conducted to evaluate compounds' pharmacological efficacy. In the topoisomerase I inhibitory test, compound 1 was most active one, 24% of inhibition at 20μM, among all the compounds but it was lower than camptothecin. Compounds 9, 11, and 13 inhibited the function of topoisomerase II more strongly than etoposide with almost same magnitude (around 90% and 30% inhibition at 100 and 20μM, respectively) which were higher than those of etoposide (72% and 18% inhibition). In the cytotoxicity test, compound 9 inhibited T47D cancer cell growth with the IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9911
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
C20H16N2O4