EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Thermodynamics of biphasic lanthanide extraction by tripodal diglycolamide: a solution calorimetry study.
PMID 27711776
Abstract
Isothermal titration calorimetry was employed for the direct measurement of the enthalpy of extraction (ΔH
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
118311
2-Decanol, 98%
C10H22O