EMAIL THIS PAGE TO A FRIEND
Oncotarget
New strategy to rescue the inhibition of osteogenesis of human bone marrow-derived mesenchymal stem cells under oxidative stress: combination of vitamin C and graphene foams.
PMID 27713129
Abstract
To rescue the oxidative stress induced inhibition of osteogenesis, vitamin C (VC) was chemically modified onto three-dimensional graphene foams (3D GFs), then their regulation on osteogenesis of human bone marrow-derived mesenchymal stem cells (BM-MSCs) was studied. Combined action of VC + GF significantly decreased H2O2-induced oxidative stress, and rescued H2O2-inhibited cell viability, differentiation and osteogenesis of BM-MSCs in vitro. Further studies revealed that Wnt pathway may be involved in this protection of osteogenesis. Furthermore, an in vivo mouse model of BM-MSCs transplantation showed that VC + GF remarkably rescued oxidative stress inhibited calcium content and bone formation. The combination of VC and GF exhibited more pronounced protective effects against oxidative stress induced inhibition of osteogenesis, compared to monotherapy of VC or GF. Our study proposed a new strategy in stem cell-based therapies for treating bone diseases.