EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
Immune Response of Healthy Adults to the Ingested Probiotic Lactobacillus casei Shirota.
PMID 27718254
Abstract
Daily ingestion of a probiotic drink containing Lactobacillus casei Shirota (LcS; 1.3 × 10