EMAIL THIS PAGE TO A FRIEND
Journal of the Formosan Medical Association = Taiwan yi zhi
High expression of mitochondrial intermembrane chaperone TIMM9 represents a negative prognostic marker in gastric cancer.
PMID 27720672
Abstract
Gastric cancer (GC) is one of the most common malignant cancers worldwide. However, little is known about the molecular process underlying this disease and its progression. This study investigated correlations between the expression of a mitochondrial inner membrane protein translocase of inner mitochondrial membrane 9 homolog (TIMM9) and various clinicopathologic parameters as well as patients' survival. Gastric tissue samples were obtained from 140 patients with GC and expression levels of TIMM9 were analyzed through immunohistochemistry. Paired t tests were used to analyze the differences in the expression levels of TIMM9 in both tumor and nontumor tissues for each patient. Two-tailed χ A borderline association was found between overexpression of TIMM9 and vascular invasion (p = 0.0887). Patients with high expression levels of TIMM9 achieved a significantly lower disease-free survival rate compared with those with low expression levels (p = 0.005). Multivariate Cox regression analysis showed that overexpression of TIMM9 was an independent prognostic marker for GC (p = 0.011). Overexpression of TIMM9 can be used as a marker to predict the outcome of patients with GC.