EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Invariant natural killer T cells play dual roles in the development of experimental autoimmune uveoretinitis.
PMID 27720708
Abstract
Experimental autoimmune uveoretinitis (EAU) represents an experimental model for human endogenous uveitis, which is caused by Th1/Th17 cell-mediated inflammation. Natural killer T (NKT) cells recognize lipid antigens and produce large amounts of cytokines upon activation. To examine the role of NKT cells in the development of uveitis, EAU was elicited by immunization with a peptide from the human interphotoreceptor retinoid-binding protein (hIRBP