EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
2-Azolylchromone Derivatives as Potent and Selective Inhibitors of Monoamine Oxidases A and B.
PMID 27725503
Abstract
A series of 2-azolylchromone derivatives were synthesized and their monoamine oxidase (MAO) A and B inhibitory activities were evaluated. Of the synthesized compounds, compounds 1b, 2b, 4a-c, 5b and 7b showed potent inhibitory activities against MAO-A (IC