EMAIL THIS PAGE TO A FRIEND
Journal of separation science
Polymer-based monolithic column with incorporated chiral metal-organic framework for enantioseparation of methyl phenyl sulfoxide using nano-liquid chromatography.
PMID 27730732
Abstract
A new approach to the preparation of enantioselective porous polymer monolithic columns with incorporated chiral metal-organic framework for nano-liquid chromatography has been developed. While no enantioseparation was achieved with monolithic poly(4-vinylpyridine-co-ethylene dimethacrylate) column, excellent separations of both enantiomers of (±)-methyl phenyl sulfoxide were achieved with its counterpart prepared after admixing metal-organic framework [Zn