EMAIL THIS PAGE TO A FRIEND
Nature communications
Polarization of M2 macrophages requires Lamtor1 that integrates cytokine and amino-acid signals.
PMID 27731330
Abstract
Macrophages play crucial roles in host defence and tissue homoeostasis, processes in which both environmental stimuli and intracellularly generated metabolites influence activation of macrophages. Activated macrophages are classified into M1 and M2 macrophages. It remains unclear how intracellular nutrition sufficiency, especially for amino acid, influences on macrophage activation. Here we show that a lysosomal adaptor protein Lamtor1, which forms an amino-acid sensing complex with lysosomal vacuolar-type H