EMAIL THIS PAGE TO A FRIEND
Endocrinology
Neuregulin 1 Regulates Proliferation of Leydig Cells to Support Spermatogenesis and Sexual Behavior in Adult Mice.
PMID 27732090
Abstract
Adult Leydig cells are derived from proliferating stem/progenitor Leydig cells in the infant testis and subsequent differentiation to steroidogenic cells in adult mice. Leydig cell proliferation in the infant testis occurs primarily in response to increased levels of LH that induce Leydig cell expression of neuregulin 1 (NRG1). Depletion of NRG1 in Nrg1 mutant mice (Nrg1
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0241 Ganirelix acetate salt, ≥95% (HPLC)
C80H113ClN18O13 · 2C2H4O2