EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Clinically Viable Gene Expression Assays with Potential for Predicting Benefit from MEK Inhibitors.
PMID 27733477
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU114431
MISSION® esiRNA, esiRNA human KRAS (esiRNA1)