EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Osthole Protects Bone Marrow-Derived Neural Stem Cells from Oxidative Damage through PI3K/Akt-1 Pathway.
PMID 27734182
Abstract
In recent years, neural stem cell (NSC) transplantation has been widely explored as a treatment for neurodegenerative diseases. NSCs are special cells that have some capacity for self-renewal and the potential to differentiate into multiple cell types. However, the inflammatory environment of diseased tissue is not conducive to the survival of transplanted cells. Osthole (Ost) is a principal bioactive component of Fructus Cnidii, Radix Angelicae Pubescentis and other traditional Chinese medicines. Ost has a wide range of pharmacological activities, such as anti-inflammation, immunomodulation, and neuroprotection. In the present study, we assessed the protective effects of Ost on bone marrow-derived-NSCs (BM-NSCs) against injury induced by hydrogen peroxide (H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4301186
Anti-Ly6a antibody produced in rabbit, affinity isolated antibody