EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Thermo-alkaline Treatment as a Practical Degradation Strategy To Reduce Indospicine Contamination in Camel Meat.
PMID 27737547
Abstract
Ingestion of indospicine-contaminated camel and horse meat has caused fatal liver injury to dogs in Australia, and it is currently not known if such contaminated meat may pose a human health risk upon dietary exposure. To date, indospicine-related research has tended to focus on analytical aspects, with little information on post-harvest management of indospicine-contaminated meat. In this study, indospicine degradation was investigated in both aqueous solution and also contaminated meat, under a range of conditions. Aqueous solutions of indospicine and indospicine-contaminated camel meat were microwaved (180 °C) or autoclaved (121 °C) with the addition of food-grade additives [0.05% (v/v) acetic acid or 0.05% (w/v) sodium bicarbonate] for 0, 15, 30, and 60 min. An aqueous sodium bicarbonate solution demonstrated the greatest efficacy in degrading indospicine, with complete degradation after 15 min of heating in a microwave or autoclave; concomitant formation of indospicine degradation products, namely, 2-aminopimelamic and 2-aminopimelic acids, was observed. Similar treatment of indospicine-contaminated camel meat with aqueous sodium bicarbonate resulted in 50% degradation after 15 min of heating in an autoclave and 100% degradation after 15 min of heating in a microwave. The results suggest that thermo-alkaline aqueous treatment has potential as a pragmatic post-harvest handling technique in reducing indospicine levels in indospicine-contaminated meat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
443751
2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%
C7H5F7O2