EMAIL THIS PAGE TO A FRIEND
Journal of virological methods
Development of a phenotypic susceptibility assay for HIV-1 integrase inhibitors.
PMID 27737783
Abstract
Phenotypic resistance analysis is an indispensable method for determination of HIV-1 resistance and cross-resistance to novel drug compounds. Since integrase inhibitors are essential components of recent antiretroviral combination therapies, phenotypic resistance data, in conjunction with the corresponding genotypes, are needed for improving rules-based and data-driven tools for resistance prediction, such as HIV-Grade and geno2pheno
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
207713
1,3-Di-o-tolylguanidine, 99%
C15H17N3