EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Effect of Tongxinluo on pulmonary hypertension and pulmonary vascular remodeling in rats exposed to a low pressure hypoxic environment.
PMID 27737815
Abstract
Tongxinluo (TXL), which is a Chinese medicine rooted from traditional used herbs, has been used in clinic to treat cardiovascular and cerebrovascular diseases. However, it remains unknown whether TXL alleviates low pressure hypoxic pulmonary hypertension. Here, we aimed to observe the influence of TXL on pulmonary hypertension in a rat model that exposed to high altitude environment characterized by low pressure hypoxia. A total of 32 male Sprague-Dawley rats were divided into four groups: control group (normal pressure and normoxia), pulmonary hypertension group (PAH, the parameter is equal to that in altitude 5000m), TXL group (rats living in environment equal to that at altitude of 5000m received TXL treatment), vardenafil group (VDNF, rats living in environment equal to that altitude of 5000m received vardenafil treatment). The high altitude environment was created in chamber by adjusting the inner pressure and oxygen content concomitantly. Before entering the chamber, the TXL group was given TXL (1.2gkg After 28 days, as compared with that in normal control group, animals living in the chamber (PAH group) showed a significant increase in mPAP( 47.5mmHg versus 18mmHg), RV/LV+IVS (0.45 versus 0.21) and MA% (78% versus 44%), respectively. Administration of TXL resulted in a significant decrease of 20mmHg in mPAP, returning of RV/LV+IVS to 0.27, and a 40% reduction in MT% compared with that in PAH group. In the VDNF group, RV/LV+IVS and MT% was 0.268 and 38.77, significantly lower than that in PAH group. While, mPAP increased by 12.5mmHg with treatment by VDNF. In contrast to the PAH group, alpha- Smooth Muscle Actin (α-SMA reduced by 19% in the TXL group (p=0.005) and 16% in the VDNF group (p=0.01). Ki67 expression in the VDNF group was significantly lower than the PAH group (P<0.01). Ki67 expression in the TXL group was significantly lower than the PAH group (P<0.01). Compared with the VDNF group, the indexes above reduced in the TXL group. Our results indicate that TXL significantly reduces pulmonary artery pressure, right ventricular hypertrophy, pulmonary small artery wall thickness, and luminal stenosis. In addition, smooth muscle proliferation markedly decreased and muscular artery decreased. However, TXL was unable to restore parameters to control levels. The automatic-adjusted low pressure hypoxic chamber was capable of establishing a pulmonary hypertension rat model at an altitude of 5000m. Compared with VDNF, TXL decreased mPAP and right ventricular hypertrophy index (RVHI) in the pulmonary hypertension rat model, and prevented vascular remodeling by reducing small pulmonary artery thickening, smooth muscle thickening and proliferation. Thus, TXL may be a potential treatment for pulmonary hypertension.