EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Urchinlike ZnS Microspheres Decorated with Nitrogen-Doped Carbon: A Superior Anode Material for Lithium and Sodium Storage.
PMID 27739614
Abstract
Urchinlike ZnS microspheres decorated with nitrogen-doped carbon (ZnS@NC) were fabricated by a facile two-step method in which urchinlike ZnS microspheres were coated with polydopamine and then calcined in an inert gas atmosphere. When employed as the anode material for lithium-ion batteries and sodium ion batteries, ZnS@NC exhibits a reversible lithium-storage capacity of 690 mAh g
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757349
Fluoroethylene carbonate, 99%
C3H3FO3