PloS one
Aqueous Extract of Gumiganghwal-tang, a Traditional Herbal Medicine, Reduces Pulmonary Fibrosis by Transforming Growth Factor-β1/Smad Signaling Pathway in Murine Model of Chronic Asthma.
PMID 27741312
Abstract
Gumiganghwal-tang is a traditional herbal prescription that is used widely for the treatment of the common cold and inflammatory diseases in Korea and other Asian countries. In this study, we investigated the protective effects of a Gumiganghwal-tang aqueous extract (GGTA) against airway inflammation and pulmonary fibrosis using a mouse model of chronic asthma. Chronic asthma was modeled in BALB/c mice via sensitization/challenge with an intraperitoneal injection of 1% ovalbumin (OVA) and inhalation of nebulized 1% OVA for 4 weeks. GGTA (100 mg/kg or 200 mg/kg) was also administered by oral gavage once a day for 4 weeks. We investigated the number of inflammatory cells, production of T-helper type 2 (Th2) cytokines, chemokine and the total transforming growth factor-β1 (TGF-β1) in bronchoalveolar lavage fluid (BALF); the levels of immunoglobulin E (IgE) in the plasma; the infiltration of inflammatory cells in lung tissue; and the expression of TGF-β1, Smad-3, and collagen in lung tissue. Our results revealed that GGTA lowered the recruitment of inflammatory cells (particularly, lymphocyte); and decreased the production of Th2 cytokines, chemokine and total TGF-β1; and attenuated the levels of total and OVA-specific IgE; and decreased the infiltration of inflammatory cells. Moreover, GGTA significantly reduced the expression of TGF-β1 and Smad-3, and lowered collagen deposition. These results indicate that GGTA reduces airway inflammation and pulmonary fibrosis by regulating Th2 cytokines production and the TGF-β1/Smad-3 pathway, thus providing a potential treatment for chronic asthma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HT110132 Eosin Y solution, alcoholic