EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
A novel L-fucose-binding lectin from Fenneropenaeus indicus induced cytotoxicity in breast cancer cells.
PMID 27742744
Abstract
Lectins are omnipresent in almost all life forms, being the proteins which specifically bind to carbohydrate moieties on the cell surface; they have been explored for their anti-tumour activities. In this study, we purified a fucose specific-lectin (IFL) from Fenneropenaeus indicus haemolymph using fucose-affinity column and characterized for its haemagglutination activity, carbohydrate specificity, dependency on cations and cytotoxicity against cancer cells. The lectin showed non-specificity against human erythrocytes. It was a Ca