EMAIL THIS PAGE TO A FRIEND
Experimental cell research
Heat stress promotes the down-regulation of IRE1α in cells: An atypical modulation of the UPR pathway.
PMID 27743894
Abstract
Heat stress induces intracellular protein denaturation and endoplasmic reticulum (ER) stress, which elicits unfolded protein response (UPR) in cells. UPR involves three ER-localized sensor proteins: the inositol-requiring protein 1α (IRE1α), the dsRNA-activated protein kinase-like ER kinase (PERK), and activating transcription factor-6 (ATF6). However, the precise mechanism by which cells deal with heat stress remains to be elucidated. We report herein that heat stress effectively activates all branches of the UPR. While splicing of the XBP-1 mRNA is usually triggered by activated IRE1α, the level of this protein was found to be decreased immediately after the occurrence of heat stress and the down-regulation gradually recovered thereafter. The protein levels of other UPR sensors or ER luminal proteins were unaffected. The down-regulation of IRE1α was independent of cellular viability. It thus appears that the heat-stress induced down-regulation of the IRE1α protein may lead to the termination of the IRE1α signaling pathway in an attempt to avoid excessive responses. We investigated the machinery causing IRE1α down-regulation and found that heat stress effectively promoted the activation of autophagy. Importantly, the inhibition of either proteasomes or autophagic flux failed to rescue the loss of IRE1α following heat stress. These collective results suggest that heat stress simultaneously activates both the UPR and autophagy, followed by the activation of a negative feedback system in UPR by modulating the responses related to the IRE1α-XBP-1 axis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9033
Thapsigargin, ≥98% (HPLC), solid film
C34H50O12