EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Bioactivation mechanism of L-thiomorpholine-3-carboxylic acid.
PMID 2774567
Abstract
L-Thiomorpholine-3-carboxylic acid (L-TMC) is a cyclized analog of S-(2-chloroethyl)-L-cysteine, which is cytotoxic in vitro and nephrotoxic in vivo. To determine whether L-TMC may play a role in S-(2-chloroethyl)-L-cysteine-induced toxicity, the cytotoxicity of L-TMC was studied in isolated rat kidney cells. L-TMC produced time- and concentration-dependent cytotoxicity. Probenecid, an inhibitor of the renal anion transport system, and L-alpha-hydroxyisocaproic acid, a substrate for L-amino acid oxidase, inhibited L-TMC-induced cytotoxicity. Rat kidney cytosol catalyzed the metabolism of L-TMC to a product absorbing at 300 nm. The increase in absorbance at 300 nm was accompanied by an increase in oxygen consumption and was inhibited by L-alpha-hydroxyisocaproic acid; moreover, the absorbance of the metabolite was quenched by addition of potassium cyanide or sodium borohydride, which indicated the formation of an imine. When L-TMC was incubated with rat kidney cytosol and sodium borodeuteride was added at the end of the incubation period, analysis by gas chromatography/mass spectrometry of the tert-butyldimethylsilyl ester of L-TMC showed the formation of [2H]TMC, indicating the intermediate formation of the imine 5,6-dihydro-2H-1,4-thiazine-3-carboxylic acid; chemically synthesized TMC imine showed similar behavior. The enzyme responsible for the metabolism of L-TMC was purified from rat kidney and was identified as L-amino acid oxidase. These observations indicate a role for L-amino acid oxidase in the bioactivation and cytotoxicity of L-TMC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219827
(S)-(−)-2-Hydroxyisocaproic acid, 98%
C6H12O3
219819
2-Hydroxyisocaproic acid, 99%
C6H12O3