EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Synthesis and kinetics investigation of meso-microporous Cu-SAPO-34 catalysts for the selective catalytic reduction of NO with ammonia.
PMID 27745671
Abstract
A series of meso-microporous Cu-SAPO-34 catalysts were successfully synthesized by a one-pot hydrothermal crystallization method, and these catalysts exhibited excellent NH
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
718203
Allyl methyl sulfone, 96%
C4H8O2S