EMAIL THIS PAGE TO A FRIEND
Frontiers in neuroscience
N-Methyl D-Aspartate Receptor Antagonist Kynurenic Acid Affects Human Cortical Development.
PMID 27746712
Abstract
Kynurenic acid (KYNA), a neuroactive metabolite of tryptophan degradation, acts as an endogenous N-methyl-D-aspartate receptor (NMDAR) antagonist. Elevated levels of KYNA have been observed in pregnant women after viral infections and are considered to play a role in neurodevelopmental disorders. However, the consequences of KYNA-induced NMDAR blockade in human cortical development still remain elusive. To study the potential impact of KYNA on human neurodevelopment, we used an