EMAIL THIS PAGE TO A FRIEND
Oncology reports
Gadolinium chloride elicits apoptosis in human osteosarcoma U-2 OS cells through extrinsic signaling, intrinsic pathway and endoplasmic reticulum stress.
PMID 27748868
Abstract
Gadolinium (Gd) compounds are important as magnetic resonance imaging (MRI) contrast agents, and are potential anticancer agents. However, no report has shown the effect of gadolinium chloride (GdCl3) on osteosarcoma inxa0vitro. The present study investigated the apoptotic mechanism of GdCl3 on human osteosarcoma U-2 OS cells. Our results indicated that GdCl3 significantly reduced cell viability of U-2 OS cells in a concentration-dependent manner. GdCl3 led to apoptotic cell shrinkage and DNA fragmentation in U-2 OS cells as revealed by morphologic changes and TUNEL staining. Colorimetric assay analyses also showed that activities of caspase-3, caspase-8, caspase-9 and caspase-4 occurred in GdCl3-treated U-2 OS cells. Pretreatment of cells with pan-caspase inhibitor (Z-VAD-FMK) and specific inhibitors of caspase-3/-8/-9 significantly reduced cell death caused by GdCl3. The increase of cytoplasmic Ca2+ level, ROS production and the decrease of mitochondria membrane potential (ΔΨm) were observed by flow cytometric analysis in U-2 OS cells after GdCl3 exposure. Western blot analyses demonstrated that the levels of Fas, FasL, cytochromexa0c, Apaf-1, GADD153 and GRP78 were upregulated in GdCl3-treated U-2 OS cells. In conclusion, death receptor, mitochondria-dependent and endoplasmic reticulum (ER) stress pathways contribute to GdCl3-induced apoptosis in U-2 OS cells. GdCl3 might have potential to be used in treatment of osteosarcoma patients.