The Journal of emergency medicine
Prognostic Value of Right Ventricular Dysfunction Markers for Serious Adverse Events in Acute Normotensive Pulmonary Embolism.
PMID 27751702
Right ventricular dysfunction (RVD) in pulmonary embolism (PE) has been associated with increased morbidity. Tools for RVD identification are not well defined. The prognostic value of RVD markers to predict serious adverse events (SAE) during hospitalization is unclear. Prospectively compare the incidence of SAE in normotensive emergency department patients with PE based upon RVD by goal-directed echocardiography (GDE), cardiac biomarkers, and right-to-left ventricle ratio by computed tomography (CT). Simplified Pulmonary Embolism Severity Index (sPESI) was calculated. Deaths and readmissions within 30xa0days were recorded. Consecutive normotensive PE patients underwent GDE focused on RVD (RV enlargement, hypokinesis, or septal bowing), serum troponin, and brain natriuretic peptide (BNP), and evaluation of the CT ventricle ratio. In-hospital SAE and complications within 30xa0days were recorded. We enrolled 123 normotensive PE patients (median age 59xa0years, 49% female). Twenty-six of 123 (26%) patients had one or more SAE. RVD was detected in 26% by GDE, in 39% by biomarkers, and in 38% with CT. In-hospital SAE included one death, six respiratory interventions, six dysrhythmias, three major bleeding episodes, and 21 hypotension episodes. Forty-one percent of patients RVD positive by GDE had SAE, compared to the 18% RVD negative by GDE. Odds ratios for GDE, CT, BNP, troponin, and sPESI for SAE were 3.2 (95% confidence interval [CI] 1.2-8.5), 2.0 (95% CI 0.8-5.1), 3.3 (95% CI 1.3-8.6), 4.2 (95% CI 1.4-13.5), and 2.9 (95% CI 1.1-8.3), respectively. Five patients had non-PE-related deaths within 30xa0days. The incidence of SAE within days of PE was significant in our cohort. Those with RVD had an increased risk of nonmortality SAE.
Related Materials
Product #
Molecular Formula
Add to Cart
(+)-Biotin 4-nitrophenyl ester, 98%