EMAIL THIS PAGE TO A FRIEND
Journal of vascular surgery
Impact of acute cerebral ischemic lesions and their volume on the revascularization outcome of symptomatic carotid stenosis.
PMID 27751736
Abstract
The influence of acute cerebral ischemic lesions (CILs) on the revascularization outcome of symptomatic carotid stenosis has been scarcely investigated in the literature. This study evaluated the effect of CILs and their volume on the results of carotid revascularization in symptomatic patients. All patients with symptomatic carotid artery stenosis who underwent carotid endarterectomy (CEA) or carotid artery stenting (CAS) between 2005 and 2014 were considered. CILs ipsilateral to the stenosis were identified in the preoperative cerebral computed tomography. The volume was quantified in mm A total of 489 symptomatic patients were treated by CEA (327 [67%]) or CAS (162 [33%]), 186 (38%) ≤2 weeks and 303 (62%) >2 weeks from symptom onset. CEA and CAS patients had statistically similar rates of stroke (3.3% vs 5.5%; P = .27) and stroke/death (3.8% vs 5.9%; P = .22). CILs were identified in 251 patients (53%) and were associated with similar stroke and stroke/death rate compared with patients without CIL (12 [4.8%] vs 8 [3.5%], P = .46; and 14 [5.6%] vs 8 [3.5%]; P = .26, respectively). The median CIL volume was 1000 mm CIL volume in symptomatic carotid stenosis seems to influence the 30-day outcome independently from the timing of carotid revascularization. A CIL volume of ≥4000 mm