EMAIL THIS PAGE TO A FRIEND
Glycoconjugate journal
Glycosaminoglycans from chicken muscular stomach or gizzard.
PMID 27752801
Abstract
Glycosaminoglycans (GAGs) were prepared from the muscular stomach or gizzard of the chicken. The content of GAGs on a dry weight basis contains 0.4 wt.% a typical value observed for a muscle tissue. The major GAG components were chondroitin-6-sulfate and chondroitin-4-sulfate (~64 %) of molecular weight 21-22 kDa. Hyaluronan (~24 %) had a molecular weight 120 kDa. Smaller amounts (12 %) of heparan sulfate was also present which was made of more highly sulfated chains of molecular weight of 21-22 kDa and a less sulfated low molecular weight (< 10 kDa) heterogeneous partially degraded heparan sulfate. Chicken gizzard represents an inexpensive and readily available source of muscle tissue-derived GAGs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
341088
1,9-Dimethyl-Methylene Blue zinc chloride double salt, Dye content 80 %
C18H22ClN3S · 0.5ZnCl2