EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Polyphosphates and Fulvates Enhance Environmental Stability of PO
PMID 27754660
Abstract
Iron oxyhydroxide nanoparticles (Fe-NPs) are natural vectors of phosphate (PO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5508
Trisodium trimetaphosphate, ≥95%
Na3O9P3