EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Silencer-of-Death Domain Mediates Acid-Induced Decrease in Cell Apoptosis in Barrett's Associated Esophageal Adenocarcinoma Cells.
PMID 27756772
Abstract
We have shown that NADPH oxidase (NOX)5-S may mediate the acid-induced decrease in cell apoptosis. However, mechanisms of NOX5-S-dependent decrease in cell apoptosis are not fully understood. In this study, we found that silencer-of-death domain (SODD) was significantly increased in esophageal adenocarcinoma (EA) tissues, EA cell lines FLO and OE33, and a dysplastic cell line CP-B. Strong SODD immunostaining was significantly higher in low-grade dysplasia (66.7%), high-grade dysplasia (81.2%), and EA (71.2%) than in Barrett's mucosa (10.5%). Acid treatment significantly increased SODD protein and mRNA expression and promoter activity in FLO cells, an increase that was significantly decreased by the knockdown of NOX5-S and nuclear factor κB (NF-κB)1 p50 with their small interfering RNAs. Similarly, acid-induced increase of SODD mRNA was blocked by knockdown of NOX5-S and p50 in a BE cell line CP-A. Overexpression of NOX5-S significantly increased SODD protein expression in FLO cells. Moreover, overexpression of NOX5-S or p50 significantly increased the SODD promoter activity and decreased the caspase 9 activity or apoptosis. NOX5-S overexpression-induced increase in SODD promoter activity was significantly decreased by knockdown of p50. In addition, acid treatment significantly decreased the caspase 9 activity, a decrease that was significantly inhibited by knockdown of SODD. Furthermore, chromatin immunoprecipitation assay showed that NF-κB1 p50 bound to SODD genomic DNA containing a NF-κB-binding element GGGGACACCCT. This binding element was further confirmed by a gel mobility shift assay. We conclude that acid-induced increase in SODD expression and decrease in cell apoptosis may depend on the activation of NOX5-S and NF-κB1 p50 in FLO cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU049781
MISSION® esiRNA, targeting human NOX5