EMAIL THIS PAGE TO A FRIEND
Oncotarget
LYN expression predicts the response to dasatinib in a subpopulation of lung adenocarcinoma patients.
PMID 27756880
Abstract
Therapies targeting SRC family kinases (SFKs) have shown efficacy in treating non-small cell lung cancer (NSCLC). However, recent clinical trials have found that the SFK inhibitor dasatinib is ineffective in some patient cohorts. Regardless, dasatinib treatment may benefit some NSCLC patient subgroups. Here, we investigated whether expression of LYN, a member of the SFK family, is associated with patient survival, the efficacy of dasatinib, and/or NSCLC cell viability. LYN expression was associated with poor overall survival in a multivariate analysis, and this association was strongest in non-smoker female patients with adenocarcinoma (ADC). In lung ADC cells, LYN expression enhanced cell proliferation, migration, and invasion. Dasatinib inhibited LYN activity and decreased cell viability in LYN-positive ADC cell lines and xenografts. Additionally, we identified the SFKs SRC and YES as candidate dasatinib targets in LYN-negative ADC cell lines. Our findings suggest that LYN is a useful prognostic marker and a selective target of dasatinib therapy in the lung ADC subpopulation especially in female non-smokers with lung ADC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS023389
Dasatinib
C22H26ClN7O2S