EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Uranium Isotopic Fractionation Induced by U(VI) Adsorption onto Common Aquifer Minerals.
PMID 27758097
Abstract
Uranium groundwater contamination due to U mining and processing affects numerous sites globally. Bioreduction of soluble, mobile U(VI) to U(IV)-bearing solids is potentially a very effective remediation strategy. Uranium isotopes (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71063
Goethite, 30-63% Fe
HFeO2