EMAIL THIS PAGE TO A FRIEND
Molecular ecology
Chromosomal inversions and ecotypic differentiation in Anopheles gambiae: the perspective from whole-genome sequencing.
PMID 27759895
Abstract
The molecular mechanisms and genetic architecture that facilitate adaptive radiation of lineages remain elusive. Polymorphic chromosomal inversions, due to their recombination-reducing effect, are proposed instruments of ecotypic differentiation. Here, we study an ecologically diversifying lineage of Anopheles gambiae, known as the Bamako chromosomal form based on its unique complement of three chromosomal inversions, to explore the impact of these inversions on ecotypic differentiation. We used pooled and individual genome sequencing of Bamako, typical (non-Bamako) An. gambiae and the sister species Anopheles coluzzii to investigate evolutionary relationships and genomewide patterns of nucleotide diversity and differentiation among lineages. Despite extensive shared polymorphism and limited differentiation from the other taxa, Bamako clusters apart from the other taxa, and forms a maximally supported clade in neighbour-joining trees based on whole-genome data (including inversions) or solely on collinear regions. Nevertheless, F
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A140
Arecaidine propargyl ester hydrobromide, solid
C10H13NO2 · HBr